Invitrogen
Manufacturer Code:MA526971
Catalog # PIMA526971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 2-106 of WTAP produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Female-lethal(2)D homolog; hFL(2)D; PNAS-132; PNAS-132 WT1-associated protein Wilms&prime tumour 1-associating protein female-lethal(2)D homolog hFL(2)D pre-mRNA-splicing regulator WTAP putative pre-mRNA splicing regulator female-lethal(2D) wilms tumor 1-associating protein; Pre-mRNA-splicing regulator WTAP; putative pre-mRNA splicing regulator female-lethal(2D); Wilms tumor 1-associating protein; Wilms' tumour 1-associating protein; WT1-associated protein
Gene Aliases: KIAA0105; Mum2; WTAP
UniProt ID: (Human) Q15007
Entrez Gene ID: (Human) 9589
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.