Invitrogen
Manufacturer Code:MA536149
Catalog # PIMA536149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JE52-72 |
Immunogen | Synthetic peptide within human C-terminal WRN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI846146 DKFZp686C2056 DNA helicase RecQ-like type 3 Exonuclease WRN RecQ protein-like 2 RecQ3 RECQL2 RECQL3 RGD1564788 similar to Werner syndrome helicase homolog Werner syndrome ATP-dependent helicase Werner syndrome ATP-dependent helicase homolog Werner syndrome homolog (human) Werner syndrome RecQ like helicase Werner syndrome RecQ helicase-like Wrn; DNA helicase, RecQ-like type 3; Exonuclease WRN; RecQ protein-like 2; RecQ3; Werner syndrome ATP-dependent helicase; Werner syndrome, RecQ helicase-like
Gene Aliases: RECQ3; RECQL2; RECQL3; WRN
UniProt ID: (Human) Q14191
Entrez Gene ID: (Human) 7486
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.