Invitrogen
Manufacturer Code:PA527319
Catalog # PIPA527319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 194 and 510 of Human WRN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA helicase, RecQ-like type 3; Exonuclease WRN; RecQ protein-like 2; RecQ3; RECQ3 RECQL2 RECQL3 DNA helicase RecQ-like type 3 Werner syndrome ATP-dependent helicase exonuclease WRN recQ protein-like 2; Werner syndrome ATP-dependent helicase; Werner syndrome, RecQ helicase-like
Gene Aliases: RECQ3; RECQL2; RECQL3; WRN
UniProt ID: (Human) Q14191
Entrez Gene ID: (Human) 7486
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.