Novus Biologicals
Manufacturer Code:NBP220914
Catalog # NBP220914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human WRN. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686C2056 DNA helicase RecQ-like type 3 EC 3.1 EC 3.6.1 EC 3.6.4.12 Exonuclease WRN RecQ protein-like 2 RecQ3 RECQ3RECQL3 RECQL2Werner syndrome ATP-dependent helicase Werner syndrome Werner syndrome RecQ helicase-like; DNA helicase, RecQ-like type 3; Exonuclease WRN; RecQ protein-like 2; RecQ3; Werner syndrome ATP-dependent helicase; Werner syndrome, RecQ helicase-like
Gene Aliases: RECQ3; RECQL2; RECQL3; WRN
UniProt ID: (Human) Q14191
Entrez Gene ID: (Human) 7486
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.