Abnova
Manufacturer Code:H00007485B02P
Catalog # 89-002-806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | WRB (NP_004618.2 1 a.a. ~ 174 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human WRB protein.
For Research Use Only
Protein Aliases: CHD5; Congenital heart disease 5 protein; Guided entry of tail-anchored proteins factor 1; Tail-anchored protein insertion receptor WRB; Tryptophan-rich basic protein
Gene Aliases: CHD5; GET1; WRB
UniProt ID: (Human) O00258
Entrez Gene ID: (Human) 7485
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.