Invitrogen
Manufacturer Code:PA5106452
Catalog # PIPA5106452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human WRAP53 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC021790 DKCB3 TCAB1 Telomerase Cajal body protein 1 WD repeat containing antisense to TP53 WD repeat containing antisense to TP53 WD repeat containing antisense to Trp53 WD repeat domain 79 WD repeat-containing protein 79 WD repeat-containing protein WRAP53 WD40 protein Wrap53 WD40 repeat-containing protein antisense to TP53 WD40 repeat-containing protein antisense to TP53 gene WD40 repeat-containing protein antisense to TP53 gene homolog WD40 repeat-containing protein encoded by RNA antisense to p53 WD40 repeat-containing protein encoding RNA antisense to p53 Wdr79 Wrap53 WRAP53beta; Telomerase Cajal body protein 1; WD repeat containing, antisense to TP53; WD repeat domain 79; WD repeat-containing protein 79; WD repeat-containing protein WRAP53; WD40 protein Wrap53; WD40 repeat-containing protein antisense to TP53; WD40 repeat-containing protein antisense to TP53 gene; WD40 repeat-containing protein antisense to TP53 gene homolog; WD40 repeat-containing protein encoded by RNA antisense to p53; WD40 repeat-containing protein encoding RNA antisense to p53; WRAP53beta
Gene Aliases: BC021790; DKCB3; TCAB1; WDR79; WRAP53
UniProt ID: (Human) B3KPR9, (Rat) Q5XII5, (Mouse) Q8VC51
Entrez Gene ID: (Human) 55135, (Rat) 287432, (Mouse) 216853
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.