Invitrogen
Manufacturer Code:PA519149
Catalog # PIPA519149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KRGNKDLRARADA) corresponding to the internal amino acids of WNT9B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9B; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15; WNT9B wingless-type MMTV integration site family member 9B WNT14B WNT15 wingless-type MMTV integration site family member 15
Gene Aliases: clf; clf1; UNQ6973/PRO21956; wnt-14b; wnt-15; WNT14B; WNT15; WNT9B
UniProt ID: (Human) O14905, (Mouse) O35468
Entrez Gene ID: (Human) 7484, (Mouse) 22412
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.