Invitrogen
Manufacturer Code:PA5116130
Catalog # PIPA5116130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human WNT9B corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: clf clf1 protein Wnt-14b Protein Wnt-15 Protein Wnt-9b UNQ6973/PRO21956 wingless-type MMTV integration site 15 wingless-type MMTV integration site 9B wingless-type MMTV integration site family member 9B wingless-type MMTV integration site family member 15 wingless-type MMTV integration site family member 9B Wnt family member 9B Wnt14b wnt-14b Wnt15 wnt-15 Wnt9b; Protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9B; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15
Gene Aliases: clf; clf1; UNQ6973/PRO21956; wnt-14b; wnt-15; WNT14B; WNT15; WNT9B
UniProt ID: (Human) O14905, (Mouse) O35468
Entrez Gene ID: (Human) 7484, (Mouse) 22412
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.