Invitrogen
Manufacturer Code:PA5117292
Catalog # PIPA5117292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human WNT9A. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC138165 MGC141991 Protein Wnt-14 Protein Wnt-9a wingless related MMTV integration site 9A wingless-type MMTV integration site 14 wingless-type MMTV integration site 9A wingless-type MMTV integration site family member 9A wingless-type MMTV integration site family member 14 wingless-type MMTV integration site family member 9A Wnt family member 9A WNT14 wnt-14 WNT9A; Protein Wnt-14; Protein Wnt-9a; wingless related MMTV integration site 9A; wingless-type MMTV integration site 14; wingless-type MMTV integration site 9A; wnt-14
Gene Aliases: Wnt14; Wnt9a
UniProt ID: (Mouse) Q8R5M2
Entrez Gene ID: (Rat) 287357, (Mouse) 216795
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.