Invitrogen
Manufacturer Code:PA568600
Catalog # PIPA568600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human WNT8B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-8b; Protein Wnt-8b wingless-type MMTV integration site family member 8B; wingless related MMTV integration site 8b; wingless-type MMTV integration site family member 8B; wingless-type MMTV integration site family, member 8B; wnt8b
Gene Aliases: wnt-8b; WNT8B
UniProt ID: (Human) Q93098, (Zebrafish) P51029
Entrez Gene ID: (Human) 7479, (Horse) 100070360, (Rabbit) 100357933, (Dog) 486841, (Guinea pig) 100724403, (Mouse) 22423, (Rat) 293990, (Zebrafish) 30144
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.