Invitrogen
Manufacturer Code:PA570530
Catalog # PIPA570530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human WNT8A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-8a; Protein Wnt-8d; Stimulated by retinoic acid gene 11 protein; wingless-related MMTV integration site 8A; wingless-related MMTV integration site 8D; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; wnt-8; WNT8A Protein Wnt-8d WNT8D Protein Wnt-8a; WNT8d
Gene Aliases: Stra11; Wnt-8A; Wnt-8D; WNT8A; WNT8D
UniProt ID: (Human) Q9H1J5, (Mouse) Q64527
Entrez Gene ID: (Bovine) 540199, (Horse) 100072609, (Pig) 100739710, (Human) 7478, (Rabbit) 100342870, (Dog) 100855884, (Mouse) 20890, (Rat) 291678, (Guinea pig) 100725534
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.