Abnova
Manufacturer Code:PAB28354
Catalog # 89-129-856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids at internal region of human WNT8A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyconal antibody raised against synthetic peptide of WNT8A.
For Research Use Only
Protein Aliases: Protein Wnt-8a; Protein Wnt-8d; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; WNT8D
Gene Aliases: WNT8A; WNT8D
UniProt ID: (Human) Q9H1J5
Entrez Gene ID: (Human) 7478
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.