Invitrogen
Manufacturer Code:PA547430
Catalog # PIPA547430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human Wnt-7a Leu32-Glu72 Thr216-Arg292 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-7a; protein Wnt-7a proto-oncogene Wnt7a protein protein Wnt-7b proto-oncogene Wnt7b protein; Protein Wnt-7b; proto-oncogene Wnt7a protein; wingless-type MMTV integration site family member 7A; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7A; wingless-type MMTV integration site family, member 7B
Gene Aliases: WNT7A; WNT7B
UniProt ID: (Human) O00755, (Human) P56706
Entrez Gene ID: (Human) 7476, (Human) 7477
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.