Proteintech
Manufacturer Code:106051AP150UL
Catalog # 50-172-6443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WNT7A/B Fusion Protein Ag0874 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: postaxial hemimelia; Protein Wnt 7a WNT7A; Protein Wnt-7a; Protein Wnt-7b; proto-oncogene Wnt7a protein; wingless-related MMTV integration site 7A; wingless-related MMTV integration site 7B; wingless-type MMTV integration site 7A; wingless-type MMTV integration site family member 7A; wingless-type MMTV integration site family member 7B; wingless-type MMTV integration site family, member 7A; wingless-type MMTV integration site family, member 7B
Gene Aliases: AI849442; px; tw; Wnt-7a; Wnt-7b; WNT7A; WNT7B
UniProt ID: (Human) O00755, (Human) P56706, (Mouse) P24383, (Mouse) P28047
Entrez Gene ID: (Human) 7476, (Human) 7477, (Rat) 114850, (Mouse) 22421, (Rat) 315196, (Mouse) 22422
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.