Proteintech
Manufacturer Code:551841AP150UL
Catalog # 50-173-6042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hWNT5A Protein Wnt 5a WNT5A WNT5A B WNT5A/B WNT5B; Protein Wnt-5a; Protein Wnt-5b; wingless-related MMTV integration site 5A; wingless-related MMTV integration site 5B; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family member 5B; wingless-type MMTV integration site family, member 5A; wingless-type MMTV integration site family, member 5B; WNT-5A protein; WNT-5B protein
Gene Aliases: 8030457G12Rik; AW545702; hWNT5A; Wnt-5a; Wnt-5b; WNT5A; WNT5B
UniProt ID: (Human) P41221, (Human) Q9H1J7, (Mouse) P22725, (Mouse) P22726
Entrez Gene ID: (Human) 7474, (Human) 81029, (Mouse) 22418, (Mouse) 22419
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.