Invitrogen
Manufacturer Code:PA585302
Catalog # PIPA585302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human WNT5A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hWNT5A antibody WNT5A antibody protein Wnt-5a antibody WNT-5A protein antibody wingless-type MMTV integration site family member 5A antibody; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; WNT-5A protein
Gene Aliases: 8030457G12Rik; hWNT5A; Wnt-5a; WNT5A
UniProt ID: (Human) P41221, (Mouse) P22725
Entrez Gene ID: (Human) 7474, (Rat) 64566, (Mouse) 22418
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.