Invitrogen
Manufacturer Code:PA573004
Catalog # PIPA573004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 190-230 of human Wnt5A was used as the immunogen for this antibody GenBank no NP&lowbar003383.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OTTHUMP00000212837 OTTHUMP00000212861 WNT-5A protein protein Wnt-5a; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; WNT-5A protein
Gene Aliases: 8030457G12Rik; hWNT5A; Wnt-5a; WNT5A
UniProt ID: (Human) P41221, (Mouse) P22725
Entrez Gene ID: (Bovine) 530005, (Human) 7474, (Rat) 64566, (Mouse) 22418
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.