Abnova
Manufacturer Code:H00007474K
Catalog # 89-203-475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human WNT5A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human WNT5A peptide using ARM Technology.
For Research Use Only
Protein Aliases: hWNT5A; Protein Wnt-5a; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; WNT-5A protein
Gene Aliases: hWNT5A; WNT5A
UniProt ID: (Human) P41221
Entrez Gene ID: (Human) 7474
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.