Abnova
Manufacturer Code:PAB9584
Catalog # 89-134-250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 10 of human WNT4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of WNT4.
For Research Use Only
Protein Aliases: Protein Wnt-4; SERKAL; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4; WNT-4
Gene Aliases: SERKAL; UNQ426/PRO864; WNT-4; WNT4
UniProt ID: (Human) P56705
Entrez Gene ID: (Human) 54361
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.