Invitrogen
Manufacturer Code:PA587468
Catalog # PIPA587468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-352 of human WNT3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-3a; vestigial tail; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3A
Gene Aliases: vt; Wnt-3a; WNT3A
UniProt ID: (Human) P56704, (Mouse) P27467
Entrez Gene ID: (Human) 89780, (Rat) 303181, (Mouse) 22416
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.