Invitrogen
Manufacturer Code:PA544946
Catalog # PIPA544946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human WNT3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-3a; vestigial tail; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; Wingless-type MMTV integration site family member 3A MGC119418 MGC119419 MGC119420; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; wnt3 like
Gene Aliases: vt; Wnt-3a; wnt3; wnt3 l; WNT3A; wnt3l; wnt[a]; Zwnt[a]
UniProt ID: (Human) P56704, (Mouse) P27467
Entrez Gene ID: (Pig) 100514950, (Horse) 100061145, (Dog) 482208, (Rabbit) 100354583, (Human) 89780, (Rat) 303181, (Mouse) 22416, (Zebrafish) 60632
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.