Invitrogen
Manufacturer Code:PA518516
Catalog # PIPA518516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CGRGHNTRTEKRKEK) corresponding to the internal amino acids of WNT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Proto-oncogene Int-4; Proto-oncogene Int-4 homolog; proto-oncogene protein Wnt-3; Proto-oncogene Wnt-3; wingless-related MMTV integration site 3; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; WNT-3 proto-oncogene protein; WNT3 wingless-type MMTV integration site family member 3 INT4 MGC131950 MGC138321 MGC138323 WNT-3 proto-oncogene protein precursor
Gene Aliases: Int-4; INT4; TETAMS; Wnt-3; WNT3
UniProt ID: (Human) P56703, (Mouse) P17553
Entrez Gene ID: (Human) 7473, (Mouse) 22415
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.