Invitrogen
Manufacturer Code:PA578424
Catalog # PIPA578424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human WNT2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Int-1-like protein 1; INT-1-related protein; INT1L1 IRP WNT2 int-1-like protein 1 protein Wnt-2 Int-1-related protein secreted growth factor wingless-type MMTV integration site family member 2; IRP; Protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site family member 2
Gene Aliases: 2610510E18Rik; INT1L1; IRP; Mirp; Wnt-2; WNT2; Wnt2a
UniProt ID: (Human) P09544, (Mouse) P21552
Entrez Gene ID: (Human) 7472, (Mouse) 22413
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.