Invitrogen
Manufacturer Code:PA576844
Catalog # PIPA576844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human WNT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Int 1 related protein Int-1-like protein 1 Int-1-related protein INT1L 1 INT1L1 IRP IRP protein ONCOGENE INT1 LIKE 1 Protein Wnt-2 Secreted growth factor Wingless type MMTV integration site family member 2 WNT 2 WNT2 WNT2&lowbarHUMAN; Int-1-like protein 1; Int-1-related protein; IRP; Protein Wnt-2; secreted growth factor; wingless-type MMTV integration site family member 2
Gene Aliases: INT1L1; IRP; WNT2
UniProt ID: (Human) P09544
Entrez Gene ID: (Human) 7472
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.