Invitrogen
Manufacturer Code:PA568544
Catalog # PIPA568544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human WNT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: etID309852.18; Int-1-like protein 1; INT-1-related protein; IRP; Protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; WNT2 IRP Int-1-like protein 1 Int-1-related protein Protein Wnt-2 Secreted growth factor INT1L1
Gene Aliases: 2610510E18Rik; INT1L1; IRP; Mirp; Wnt; Wnt-2; WNT2; Wnt2a; ZfWnt2
UniProt ID: (Human) P09544, (Sheep) Q09YI4, (Bovine) A4D7S0, (Horse) Q2QLA5, (Rabbit) Q09YN1, (Mouse) P21552, (Zebrafish) Q92048
Entrez Gene ID: (Human) 7472, (Sheep) 100126577, (Bovine) 493729, (Horse) 100056092, (Rabbit) 100126569, (Dog) 612141, (Guinea pig) 100719346, (Mouse) 22413, (Rat) 114487, (Zebrafish) 30127
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.