Invitrogen
Manufacturer Code:PA578493
Catalog # PIPA578493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human WNT16. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-16; wingless-related MMTV integration site 16; wingless-type MMTV integration site family member 16; wingless-type MMTV integration site family member 16b; wingless-type MMTV integration site family, member 16
Gene Aliases: E130309I19Rik; WNT16
UniProt ID: (Human) Q9UBV4, (Mouse) Q9QYS1
Entrez Gene ID: (Human) 51384, (Mouse) 93735
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.