Invitrogen
Manufacturer Code:PA572461
Catalog # PIPA572461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 193-222 amino acids from the Central region of human WNT10B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wnt-10b; Protein Wnt-12; wingless related MMTV integration site 10b; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; WNT-10B protein; wnt-12; WNT10B Protein Wnt-10b SHFM6 WNT-12 WNT-10B protein WNT12 Protein Wnt-12
Gene Aliases: SHFM6; Wnt-10b; WNT-12; Wnt10; WNT10B; WNT12
UniProt ID: (Human) O00744, (Mouse) P48614
Entrez Gene ID: (Human) 7480, (Mouse) 22410
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.