Invitrogen
Manufacturer Code:PA532641
Catalog # PIPA532641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from N-terminus of human WNT-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: INT1 proto-oncogene Int-1 homolog proto-oncogene Wnt-1 wingless-type MMTV integration site family member 1 oncogene INT1; Proto-oncogene Int-1; Proto-oncogene Int-1 homolog; proto-oncogene protein Wnt-1; Proto-oncogene Wnt-1; wingless-related MMTV integration site 1; wingless-type MMTV integration site family member 1; wingless-type MMTV integration site family, member 1 (oncogene INT1)
Gene Aliases: BMND16; Int-1; INT1; OI15; sw; swaying; Wnt-1; WNT1
UniProt ID: (Human) P04628, (Mouse) P04426
Entrez Gene ID: (Human) 7471, (Mouse) 22408
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.