Novus Biologicals
Manufacturer Code:NB600284F
Catalog # NB600284F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to a region of the human WNK4 protein between residues 200-300. [UniProt# Q96J92] |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 PHA2B PRKWNK4protein kinase lysine deficient 4 Protein kinase lysine-deficient 4 Protein kinase with no lysine 4 serine/threonine-protein kinase WNK4 WNK lysine deficient protein kinase 4; Protein kinase lysine-deficient 4; Protein kinase with no lysine 4; Serine/threonine-protein kinase WNK4
Gene Aliases: PHA2B; PRKWNK4; WNK4
UniProt ID: (Human) Q96J92
Entrez Gene ID: (Human) 65266
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.