Proteintech
Manufacturer Code:223261AP150UL
Catalog # 50-173-3829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WNK4 Fusion Protein Ag17773 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PHA2B PRKWNK4 WNK4; Protein kinase lysine-deficient 4; Protein kinase with no lysine 4; protein kinase, lysine deficient 4; Serine/threonine-protein kinase WNK4
Gene Aliases: 2010002J11Rik; PHA2B; PRKWNK4; WNK4
UniProt ID: (Human) Q96J92, (Mouse) Q80UE6
Entrez Gene ID: (Human) 65266, (Mouse) 69847
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.