Invitrogen
Manufacturer Code:PA576762
Catalog # PIPA576762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human WNK3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1566PRKWNK 3PRKWNK3Protein kinase lysine deficient 3Protein kinase lysine-deficient 3Protein kinase with no lysine 3Serine/threonine protein kinase WNK3Serine/threonine-protein kinase WNK3WNK 3WNK lysine deficient protein kinase 3Wnk3WNK3&lowbarHUMAN; Protein kinase lysine-deficient 3; Protein kinase with no lysine 3; Serine/threonine-protein kinase WNK3; WNK lysine deficient protein kinase 3, pseudogene
Gene Aliases: KIAA1566; PRKWNK3; RGD1563131; WNK3; Wnk3-ps
UniProt ID: (Human) Q9BYP7, (Mouse) Q80XP9
Entrez Gene ID: (Human) 65267, (Mouse) 279561, (Rat) 317420
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.