Invitrogen
Manufacturer Code:PA5102806
Catalog # PIPA5102806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human WNK3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1566 PRKWNK 3 PRKWNK3 protein kinase lysine-deficient 3 Protein kinase with no lysine 3 RGD1563131 serine/threonine-protein kinase WNK3 WNK 3 WNK lysine deficient protein kinase 3 WNK lysine deficient protein kinase 3 pseudogene Wnk3 Wnk3-ps; Protein kinase lysine-deficient 3; Protein kinase with no lysine 3; Serine/threonine-protein kinase WNK3; WNK lysine deficient protein kinase 3, pseudogene
Gene Aliases: KIAA1566; PRKWNK3; RGD1563131; WNK3; Wnk3-ps
UniProt ID: (Human) Q9BYP7, (Mouse) Q80XP9
Entrez Gene ID: (Human) 65267, (Mouse) 279561, (Rat) 317420
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.