Invitrogen
Manufacturer Code:PA554636
Catalog # PIPA554636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WIZ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Wiz; widely-interspaced zinc finger motifs; Widely-interspaced zinc finger-containing protein; WIZ zinc finger; Zinc finger protein 803; ZNF803 WIZ zinc finger protein Wiz widely-interspaced zinc finger motifs widely-interspaced zinc finger-containing protein zinc finger protein 803
Gene Aliases: WIZ; ZNF803
UniProt ID: (Human) O95785
Entrez Gene ID: (Human) 58525
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.