Novus Biologicals
Manufacturer Code:NBP188878
Catalog # NBP188878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:HQDRAFATARLNFSGQRNICTLSTIQKLPRLLVASSSGHLYMYNLDPQDGGECVLIKTHSLLGSGTTEENKENDLRPSLPQSYAATVARPSASSASTVPGYSEDGGALRGEVIPEHEFATGPVCLDDENEFPPIILCRG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Atg18 Atg18 protein homolog ATG18A FLJ10055 WD repeat domain phosphoinositide-interacting protein 1 WD repeat domain phosphoinositide interacting 1 WD40 repeat protein interacting with phosphoinositides of 49 kDa WD40 repeat protein Interacting with phosphoInositides of 49kDa WIPI 49 kDa WIPI-1 WIPI-1 alpha WIPI49ATG18; Atg18 protein homolog; WD repeat domain phosphoinositide-interacting protein 1; WD40 repeat protein interacting with phosphoinositides of 49 kDa; WIPI 49 kDa; WIPI-1; WIPI-1 alpha
Gene Aliases: ATG18; ATG18A; WIPI1; WIPI49
UniProt ID: (Human) Q5MNZ9
Entrez Gene ID: (Human) 55062
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.