Abnova
Manufacturer Code:PAB27515
Catalog # 89-129-024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human WIPF3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant WIPF3.
For Research Use Only
Protein Aliases: Corticosteroids and regional expression protein 16 homolog; CR16; FLJ36931; WAS/WASL interacting protein family, member 3; WAS/WASL-interacting protein family member 3
Gene Aliases: CR16; WIPF3
UniProt ID: (Human) A6NGB9
Entrez Gene ID: (Human) 644150
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.