Invitrogen
Manufacturer Code:PA555086
Catalog # PIPA555086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WIPF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: WAS/WASL interacting protein family, member 2; WAS/WASL-interacting protein family member 2; WASP-binding protein; WASP-interacting protein-related protein; WICH WIRE WAS/WASL-interacting protein family member 2 WASP-binding protein WASP-interacting protein-related protein WIP- and CR16-homologous protein WIP-related protein; WIP- and CR16-homologous protein; WIP-related protein
Gene Aliases: 1110014J05Rik; 5730509C05Rik; AA407487; Gm1176; PP10631; RGD1561080; WICH; WIPF2; WIRE
UniProt ID: (Human) Q8TF74, (Mouse) Q6PEV3
Entrez Gene ID: (Human) 147179, (Mouse) 68524, (Rat) 360620
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.