Invitrogen
Manufacturer Code:PA524457
Catalog # PIPA524457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 392-419 amino acids from the C-terminal region of human WIPF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: WAS/WASL interacting protein family, member 2; WAS/WASL-interacting protein family member 2; WASP-binding protein; WASP-interacting protein-related protein; WICH WIRE WAS/WASL-interacting protein family member 2 WASP-binding protein WASP-interacting protein-related protein WIP- and CR16-homologous protein WIP-related protein; WIP- and CR16-homologous protein; WIP-related protein
Gene Aliases: PP10631; WICH; WIPF2; WIRE
UniProt ID: (Human) Q8TF74
Entrez Gene ID: (Human) 147179
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.