Invitrogen
Manufacturer Code:PA551995
Catalog # PIPA551995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WIPF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein PRPL-2; testicular tissue protein Li 226; WAS/WASL interacting protein family, member 1; WAS/WASL-interacting protein family member 1; WASP-interacting protein; WIPF1 WAS/WASL interacting protein family member 1 MGC111041 PRPL-2 WASPIP WIP Wiskott-Aldrich syndrome protein interacting protein; Wiskott-Aldrich syndrome protein interacting protein; Wiskott-Aldrich syndrome protein-interacting protein
Gene Aliases: PRPL-2; WAS2; WASPIP; WIP; WIPF1
UniProt ID: (Human) O43516
Entrez Gene ID: (Human) 7456
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.