Invitrogen
Manufacturer Code:PA570518
Catalog # PIPA570518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human WHSC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10442 FLJ25112 NELF-A NELFA P/OKcl.15; mWHSC2; Negative elongation factor A; NELF-A; Wolf-Hirschhorn syndrome candidate 2; Wolf-Hirschhorn syndrome candidate 2 homolog; Wolf-Hirschhorn syndrome candidate 2 protein
Gene Aliases: NELF-A; NELFA; P/OKcl.15; WHSC2; Whsc2h
UniProt ID: (Human) Q9H3P2, (Mouse) Q8BG30
Entrez Gene ID: (Human) 7469, (Pig) 100514855, (Bovine) 789389, (Mouse) 24116, (Rat) 305455
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.