Abnova
Manufacturer Code:PAB30540
Catalog # 89-032-881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human WHSC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ23286; Histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; KIAA1090; MGC176638; MMSET; multiple myeloma SET domain containing protein type III; Multiple myeloma SET domain-containing protein; NSD2; nuclear receptor binding SET domain protein 2; Nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; REIIBP; trithorax/ash1-related protein 5; TRX5; WHS; Wolf-Hirschhorn syndrome candidate 1 protein
Gene Aliases: KIAA1090; MMSET; NSD2; REIIBP; TRX5; WHS; WHSC1
UniProt ID: (Human) O96028
Entrez Gene ID: (Human) 7468
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.