Proteintech
Manufacturer Code:115581AP150UL
Catalog # 50-172-7265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WFS1 Fusion Protein Ag2114 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: WFRS WFS WFS1 Wolfram syndrome 1 (wolframin) WOLFRAMIN; Wolfram syndrome 1 (wolframin); Wolfram syndrome 1 protein homolog; Wolframin
Gene Aliases: AI481085; CTRCT41; WFRS; WFS; WFS1; WFSL; wolframin
UniProt ID: (Human) O76024, (Mouse) P56695
Entrez Gene ID: (Human) 7466, (Mouse) 22393
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.