Invitrogen
Manufacturer Code:PA580227
Catalog # PIPA580227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat HE4 recombit protein (Position: E31-F168). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epididymal secretory protein 4; RE4; WAP domain-containing protein HE4; WAP four-disulfide core domain protein 2; WAP four-disulfide core domain protein 2 Epididymal secretory protein 4 RE4 Wfdc2
Gene Aliases: 1600023A02Rik; HE4; re4; WAP5; Wfdc2
UniProt ID: (Rat) Q8CHN3, (Mouse) Q9DAU7
Entrez Gene ID: (Rat) 286888, (Mouse) 67701
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.