Invitrogen
Manufacturer Code:PA580226
Catalog # PIPA580226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human HE4 recombit protein (Position: E31-F124). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymal protein 4; Epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; Major epididymis-specific protein E4; Putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain protein 2; WAP four-disulfide core domain protein 2 Epididymal secretory protein E4 Major epididymis-specific protein E4 Putative protease inhibitor WAP5 WFDC2 HE4 WAP5
Gene Aliases: 1600023A02Rik; dJ461P17.6; EDDM4; HE4; re4; WAP5; WFDC2
UniProt ID: (Human) Q14508, (Rat) Q8CHN3, (Mouse) Q9DAU7
Entrez Gene ID: (Human) 10406, (Rat) 286888, (Mouse) 67701
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.