Proteintech
Manufacturer Code:132041AP150UL
Catalog # 50-172-8568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WFDC1 Fusion Protein Ag3960 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Prostate stromal protein ps20; Prostate stromal protein ps20 PS20 ps20 growth inhibitor WFDC1; ps20 growth inhibitor; WAP four-disulfide core domain 1 homolog; WAP four-disulfide core domain protein 1
Gene Aliases: PS20; WFDC1
UniProt ID: (Human) Q9HC57
Entrez Gene ID: (Human) 58189
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.