Proteintech
Manufacturer Code:551191AP150UL
Catalog # 50-173-5997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: WEE1 homolog 2 (S. pombe) Wee1 like protein kinase 1B Wee1 like protein kinase 2 WEE1B Wee1B kinase WEE2; Wee1-like protein kinase 1B; Wee1-like protein kinase 2; Wee1B kinase
Gene Aliases: WEE1B; WEE2
UniProt ID: (Human) P0C1S8
Entrez Gene ID: (Human) 494551
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.