Abnova
Manufacturer Code:H00023038K
Catalog # 89-207-104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human WDTC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human WDTC1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: adipose homolog; ADP KIAA1037; DDB1 and CUL4 associated factor 9; WD and tetratricopeptide repeats protein 1
Gene Aliases: ADP; DCAF9; KIAA1037; WDTC1
UniProt ID: (Human) Q8N5D0
Entrez Gene ID: (Human) 23038
Molecular Function: G-protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.