Proteintech
Manufacturer Code:255401AP150UL
Catalog # 50-173-4953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WDR92 Fusion Protein Ag22181 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: monad; testicular secretory protein Li 67; WD repeat domain 92 WDR92; WD repeat-containing protein 92; WD repeat-containing protein Monad
Gene Aliases: AI553587; HZGJ; WDR92
UniProt ID: (Human) Q96MX6, (Mouse) Q8BGF3
Entrez Gene ID: (Human) 116143, (Mouse) 103784
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.