Proteintech
Manufacturer Code:276281AP150UL
Catalog # 50-173-5719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WDR8 Fusion Protein Ag26267 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: WD repeat containing protein 8 WD repeat domain 8 WD repeat-containing protein WRAP73 WDR8 WRAP73; WD repeat domain 8; WD repeat-containing protein 8; WD repeat-containing protein antisense to TP73 gene; WD repeat-containing protein WRAP73
Gene Aliases: WDR8; WRAP73
UniProt ID: (Human) Q5T0D6
Entrez Gene ID: (Human) 49856
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.