Novus Biologicals
Manufacturer Code:NBP274867FR
Catalog # NBP274867FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 114-385 of human WDR74 (NP_060563) produced in E.coli. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ10439 FLJ21730 NOP seven-associated protein 1 NSA1 WD repeat domain 74 WD repeat-containing protein 74; NOP seven-associated protein 1; WD repeat-containing protein 74
Gene Aliases: NSA1; WDR74
UniProt ID: (Human) Q6RFH5
Entrez Gene ID: (Human) 54663
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.