Invitrogen
Manufacturer Code:PA520924
Catalog # PIPA520924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the carboxy terminus of human NSA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NOP seven-associated protein 1; NOP seven-associated protein 1 WD repeat-containing protein 74; WD repeat-containing protein 74
Gene Aliases: 5730436H21Rik; AA407588; NSA1; WDR74
UniProt ID: (Human) Q6RFH5, (Mouse) Q8VCG3
Entrez Gene ID: (Human) 54663, (Rat) 690229, (Mouse) 107071
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.